Assay Details
Target Gene Details
Entrez Gene ID: | 19876 |
Gene Name: | roundabout homolog 1 (Drosophila) |
Gene Aliases: |
AW494633, AW742721, DUTT1, Gm310 |
Location: |
Chr.16:72027553-73047491 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Robo1 | NM_019413.2 | NP_062286.2 | ||
| AK138485.1 | BAE23679.1 | |||
| AK142047.1 | BAE24924.1 | |||
| Y17793.1 | CAA76850.1 |
More Information
Set Membership: |
|

Genomic Map