Assay Details
Target Gene Details
Entrez Gene ID: | 73333 |
Gene Name: | solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31 |
Gene Aliases: |
1700034J06Rik, Ant4, Sfec |
Location: |
Chr.3:40708871-40726094 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a31 | NM_178386.3 | NP_848473.2 | ||
| AK006599.1 | ||||
| AK132722.1 | BAE21321.1 | |||
| AY550241.1 | ||||
| BC050810.1 | AAH50810.1 | |||
| BC139243.1 | ||||
| BC139245.1 |
More Information
Set Membership: |
|

Genomic Map