Assay Details
Target Gene Details
Entrez Gene ID: | 21949 |
Gene Name: | tumor necrosis factor (ligand) superfamily, member 8 |
Gene Aliases: |
CD153, CD30LG, Cd30l |
Location: |
Chr.4:63832820-63861681 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsf8 | NM_009403.3 | NP_033429.1 | ||
| AK030916.1 | BAC27182.1 | |||
| AK157270.1 | BAE34022.1 | |||
| BC117093.1 | ||||
| BC117097.1 | ||||
| L09754.1 | AAA74595.1 |
More Information
Set Membership: |
|

Genomic Map