Assay Details
Target Gene Details
Entrez Gene ID: | 257630 |
Gene Name: | interleukin 17F |
Gene Aliases: |
C87042, IL-17F |
Location: |
Chr.1:20777146-20785274 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il17f | NM_145856.2 | NP_665855.2 | ||
| AB116259.1 | BAC81535.1 | |||
| AF458064.1 | AAM77568.1 | |||
| AY380822.1 | AAQ88439.1 |
More Information
Set Membership: |
|

Genomic Map