Assay Details
Target Gene Details
Entrez Gene ID: | 282663 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 1b |
Gene Aliases: |
6330533H24Rik, EIB, Serpin1b1, ovalbumin |
Location: |
Chr.13:33078504-33094380 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb1b | NM_173052.2 | NP_766640.1 | ||
| AF426025.1 | AAL57487.1 | |||
| AK018226.1 | ||||
| BC030354.1 | AAH30354.1 | |||
| BY109795.1 |
More Information
Set Membership: |
|

Genomic Map