Assay Details
Target Gene Details
Entrez Gene ID: | 26451 |
Gene Name: | ribosomal protein L27A |
Gene Aliases: |
L27', L27A, L29 |
Location: |
Chr.7:109519195-109522369 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl27a | NM_011975.3 | NP_036105.2 | ||
| AK007837.1 | BAB25295.1 | |||
| AK008531.1 | BAB25724.1 | |||
| AK010286.1 | BAB26822.1 | |||
| AK011361.1 | ||||
| AK138110.1 | ||||
| AK150907.1 | BAE29949.1 | |||
| AK167792.1 | BAE39822.1 | |||
| AK167915.1 | BAE39922.1 | |||
| BC086939.1 | AAH86939.1 | |||
| CJ072009.1 | ||||
| X05021.1 | CAA28678.1 |
More Information
Set Membership: |
|

Genomic Map