Assay Details
Target Gene Details
Entrez Gene ID: | 19253 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 18 |
Gene Aliases: |
FLP1, HSCF, PTP-HSCF, PTP-K1, Ptpk1 |
Location: |
Chr.1:34459746-34473784 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn18 | NM_011206.2 | NP_035336.2 | ||
| AK144353.1 | BAE25845.1 | |||
| BC008512.1 | AAH08512.1 | |||
| U35124.1 | AAB82736.1 | |||
| U49853.1 | AAB18623.1 | |||
| U52523.1 | AAC52991.1 |
More Information
Set Membership: |
|

Genomic Map