Assay Details
Target Gene Details
Entrez Gene ID: | 13449 |
Gene Name: | docking protein 2 |
Gene Aliases: |
DokR, Frip |
Location: |
Chr.14:70764528-70778494 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dok2 | NM_010071.2 | NP_034201.1 | ||
| AF030627.1 | AAC31315.1 | |||
| AF035117.1 | AAC13266.1 | |||
| AF059583.1 | AAC78606.1 | |||
| AK155579.1 | BAE33334.1 | |||
| AK158246.1 | BAE34423.1 | |||
| AK159466.1 | BAE35107.1 | |||
| AK159472.1 | BAE35113.1 | |||
| BC004590.1 | AAH04590.1 |
More Information
Set Membership: |
|

Genomic Map