Assay Details
Target Gene Details
Entrez Gene ID: | 78753 |
Gene Name: | lipase, family member M |
Gene Aliases: |
4632427C23Rik, AW227514, Lipl3 |
Location: |
Chr.19:34100943-34122687 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lipm | NM_023903.1 | NP_076392.1 | ||
| AK019504.1 | ||||
| AK037091.1 | BAC29699.1 | |||
| AK037214.1 | BAC29757.1 | |||
| AK085719.1 | BAC39517.1 | |||
| BC031933.1 | AAH31933.1 |
More Information
Set Membership: |
|

Genomic Map