Assay Details
Target Gene Details
Entrez Gene ID: | 216233 |
Gene Name: | suppressor of cytokine signaling 2 |
Gene Aliases: |
8030460M17, AI527257, AW108012, CIS2, Cish2, D130043N08Rik, JAB, SOCS-2, SSI-2, hg |
Location: |
Chr.10:95383903-95417253 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Socs2 | AK033206.1 | BAC28196.2 | ||
| AK083884.1 | 1 | 972 |
More Information
Set Membership: |
|

Genomic Map