Assay Details
Target Gene Details
Entrez Gene ID: | 20295 |
Gene Name: | chemokine (C-C motif) ligand 17 |
Gene Aliases: |
Abcd-2, Scya17, Scya17l, Tarc |
Location: |
Chr.8:94807404-94812038 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccl17 | NM_011332.3 | NP_035462.2 | ||
| AF125570.1 | AAD56600.1 | |||
| AF125571.1 | AAD56601.1 | |||
| AF125572.1 | AAD56602.1 | |||
| AJ242587.1 | CAB45256.1 | |||
| AK007663.1 | BAB25171.1 | |||
| BC028505.1 | AAH28505.1 |
More Information
Set Membership: |
|

Genomic Map