Assay Details
Target Gene Details
Entrez Gene ID: | 101513 |
Gene Name: | MOB kinase activator 2 |
Gene Aliases: |
1110017M21Rik, 2700078K21Rik, AI256456, Mmh |
Location: |
Chr.7:142008535-142061034 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mob2 | NM_028308.2 | NP_082584.1 | ||
| AF228503.1 | AAL55655.1 | |||
| AK012535.1 | BAB28303.1 | |||
| AK037090.1 | ||||
| AK135483.1 | BAE22548.1 | |||
| AK158674.1 | BAE34605.1 | |||
| BC037588.1 | AAH37588.1 |
More Information
Set Membership: |
|

Genomic Map