Assay Details
Target Gene Details
Entrez Gene ID: | 74556 |
Gene Name: | thymocyte selection associated family member 3 |
Gene Aliases: |
9130404H23Rik |
Location: |
Chr.17:66555252-66594622 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Themis3 | NM_028998.1 | NP_083274.1 | ||
| AK018663.1 | BAB31332.1 |
More Information
Set Membership: |
|

Genomic Map