Assay Details
Target Gene Details
Entrez Gene ID: | 68865 |
Gene Name: | ARV1 homolog (yeast) |
Gene Aliases: |
1110067L22Rik, AI461928, AW121084 |
Location: |
Chr.8:124722138-124740648 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arv1 | NM_026855.4 | NP_081131.1 | ||
| AK004398.1 | BAB23288.1 | |||
| BC119376.1 | ||||
| BC120631.1 |
More Information
Set Membership: |
|

Genomic Map