Assay Details
Target Gene Details
Entrez Gene ID: | 66511 |
Gene Name: | chromatin target of PRMT1 |
Gene Aliases: |
2500003M10Rik, Fop, Srag |
Location: |
Chr.3:90498539-90509888 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chtop | NM_001293776.1 | NP_001280705.1 | ||
| NM_001293777.1 | NP_001280706.1 | |||
| NM_001293778.1 | NP_001280707.1 | |||
| NM_001293779.1 | NP_001280708.1 | |||
| NM_001293780.1 | NP_001280709.1 | |||
| NM_001293781.1 | NP_001280710.1 | |||
| NM_001293782.1 | NP_001280711.1 | |||
| NM_023215.6 | NP_075704.2 | |||
| AB041654.1 | BAA95102.1 | |||
| AK004558.2 | BAB23371.1 | |||
| AK005377.1 | BAB23984.2 | |||
| AK010866.1 | BAB27232.2 | |||
| AK047914.1 | BAC33189.1 | |||
| AK077960.1 | BAC37084.1 | |||
| AK132769.1 | BAE21348.1 | |||
| AK135914.1 | ||||
| AK152492.1 | BAE31263.1 | |||
| AK190161.1 | ||||
| AK204756.1 | ||||
| AK208473.1 | ||||
| BC083333.1 | AAH83333.1 | |||
| BC108396.1 | AAI08397.1 | |||
| BC120651.1 | ||||
| BC144775.1 | ||||
| BQ746440.1 | ||||
| CX234428.1 |
More Information
Set Membership: |
|

Genomic Map