Assay Details
Target Gene Details
Entrez Gene ID: | 329731 |
Gene Name: | family with sequence similarity 19, member A3 |
Gene Aliases: |
7530404M11Rik, TAFA3, Tafa-3 |
Location: |
Chr.3:104767405-104777547 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam19a3 | NM_183224.3 | NP_899047.1 | ||
| AK078681.1 | ||||
| AY325122.1 | AAP92414.1 | |||
| BC118531.1 |
More Information
Set Membership: |
|

Genomic Map