Assay Details
Target Gene Details
Entrez Gene ID: | 67909 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 5 |
Gene Aliases: |
1700021B12Rik, Galnt15 |
Location: |
Chr.5:25181440-25220297 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galntl5 | NM_026449.3 | NP_080725.2 | ||
| AK005605.1 | BAB24147.1 | |||
| AK006193.1 | ||||
| AK016415.1 | BAB30220.1 | |||
| BC049554.1 | AAH49554.1 | |||
| BC100447.1 | AAI00448.1 | |||
| BC145757.1 |
More Information
Set Membership: |
|

Genomic Map