Assay Details
Target Gene Details
Entrez Gene ID: | 11931 |
Gene Name: | ATPase, Na+/K+ transporting, beta 1 polypeptide |
Gene Aliases: |
Atp4b, Atpb, Atpb-1, NKbeta1 |
Location: |
Chr.1:164437099-164458355 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp1b1 | NM_009721.6 | NP_033851.1 | ||
| AK005071.1 | BAB23795.1 | |||
| AK010677.1 | BAB27109.1 | |||
| AK140862.1 | BAE24500.1 | |||
| AK160405.1 | BAE35773.1 | |||
| AK161891.1 | BAE36624.1 | |||
| BC027319.1 | AAH27319.1 | |||
| BC094070.1 | AAH94070.1 | |||
| X16646.1 | CAA34639.1 | |||
| X61433.1 | CAA43675.1 |
More Information
Set Membership: |
|

Genomic Map