Assay Details
Target Gene Details
Entrez Gene ID: | 20230 |
Gene Name: | special AT-rich sequence binding protein 1 |
Gene Aliases: |
2610306G12Rik, AW413156 |
Location: |
Chr.17:51736187-51833497 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Satb1 | NM_001163630.1 | NP_001157102.1 | ||
| NM_001163631.1 | NP_001157103.1 | |||
| NM_001163632.1 | NP_001157104.1 | |||
| NM_009122.2 | NP_033148.2 | |||
| AK037740.1 | ||||
| AK088563.1 | BAC40427.1 | |||
| AK134040.1 | ||||
| AK158518.1 | BAE34542.1 | |||
| BB635001.1 | ||||
| BC011132.1 | AAH11132.1 | |||
| CT010371.1 | CAJ18578.1 | |||
| U05252.1 | AAA17372.1 |
More Information
Set Membership: |
|

Genomic Map