Assay Details
Target Gene Details
Entrez Gene ID: | 18777 |
Gene Name: | lysophospholipase 1 |
Gene Aliases: |
Pla1a |
Location: |
Chr.1:4807563-4846737 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lypla1 | NM_008866.2 | NP_032892.1 | ||
| AK002674.1 | BAB22276.1 | |||
| AK050549.1 | BAC34318.1 | |||
| AK146874.1 | BAE27497.1 | |||
| BC013536.1 | AAH13536.1 | |||
| BC052848.1 | AAH52848.1 | |||
| CT010201.1 | CAJ18409.1 | |||
| U89352.1 | AAB48627.1 |
More Information
Set Membership: |
|

Genomic Map