Assay Details
Target Gene Details
Entrez Gene ID: | 319930 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 19 |
Gene Aliases: |
C130022P09Rik, CEAL1 |
Location: |
Chr.7:19875742-19887990 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam19 | NM_177036.5 | NP_796010.1 | ||
| AK047942.1 | BAC33195.1 | |||
| AK079290.1 | BAC37596.1 | |||
| AK163794.1 | BAE37496.1 | |||
| BC107227.1 | AAI07228.1 | |||
| BC107228.2 | AAI07229.1 | |||
| BC109333.3 | AAI09334.1 |
More Information
Set Membership: |
|

Genomic Map