Assay Details
Target Gene Details
Entrez Gene ID: | 96875 |
Gene Name: | proteoglycan 4 (megakaryocyte stimulating factor, articular superficial zone protein) |
Gene Aliases: |
CACP, DOL54, JCAP, MSF, SZP, lubricin |
Location: |
Chr.1:150449412-150466221 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prg4 | NM_001110146.1 | NP_001103616.1 | ||
| NM_021400.3 | NP_067375.3 | |||
| AB034730.1 | BAA92310.1 | |||
| AK149469.1 | BAE28900.1 | |||
| BC130021.1 |
More Information
Set Membership: |
|

Genomic Map