Assay Details
Target Gene Details
Entrez Gene ID: | 20773 |
Gene Name: | serine palmitoyltransferase, long chain base subunit 2 |
Gene Aliases: |
AI173915, LCB2, mKIAA0526 |
Location: |
Chr.12:87307889-87388230 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptlc2 | NM_011479.3 | NP_035609.1 | ||
| AK044917.1 | BAC32140.1 | |||
| AK057644.1 | ||||
| AK080181.1 | BAC37842.1 | |||
| AK089019.1 | BAC40704.1 | |||
| AK133728.1 | BAE21805.1 | |||
| AK159346.1 | BAE35008.1 | |||
| AK172965.1 | BAD32243.1 | |||
| AK177584.1 | ||||
| BC003227.1 | AAH03227.1 | |||
| U27455.1 | AAC53310.1 | |||
| X95642.1 | CAA64898.1 |
More Information
Set Membership: |
|

Genomic Map