Assay Details
Target Gene Details
Entrez Gene ID: | 13360 |
Gene Name: | 7-dehydrocholesterol reductase |
Gene Aliases: |
AA409147 |
Location: |
Chr.7:143823167-143848410 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhcr7 | NM_007856.2 | NP_031882.1 | ||
| AF057368.1 | AAC40164.1 | |||
| AK146811.1 | BAE27451.1 | |||
| BC006854.1 | AAH06854.1 | |||
| CT010277.1 | CAJ18485.1 |
More Information
Set Membership: |
|

Genomic Map