Assay Details
Target Gene Details
Entrez Gene ID: | 225207 |
Gene Name: | zinc finger protein 521 |
Gene Aliases: |
B930086A16Rik, Evi3, Znf521 |
Location: |
Chr.18:13687013-13972787 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp521 | NM_145492.4 | 4 | 724 | NP_663467.1 |
| AK052655.1 | 4 | 627 | ||
| AK131130.1 | 3 | 809 | BAD21380.1 | |
| AY147406.1 | 4 | 627 | AAN39839.1 | |
| BC021376.1 | 4 | 540 | AAH21376.1 |
More Information
Set Membership: |
|

Genomic Map