Assay Details
Target Gene Details
Entrez Gene ID: | 22355 |
Gene Name: | vasoactive intestinal peptide receptor 2 |
Gene Aliases: |
VPAC2, VPAC2R, Vip2 |
Location: |
Chr.12:116077726-116159604 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vipr2 | NM_009511.2 | NP_033537.1 | ||
| AK038743.1 | ||||
| AK142357.1 | BAE25042.1 | |||
| BC138572.1 | ||||
| D28132.1 | BAA05674.1 |
More Information
Set Membership: |
|

Genomic Map