Assay Details
Target Gene Details
Entrez Gene ID: | 12667 |
Gene Name: | chordin |
Gene Aliases: |
Chd |
Location: |
Chr.16:20731417-20742384 on Build GRCm38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrd | NM_001278041.1 | NP_001264970.1 | ||
| NM_009893.2 | NP_034023.1 | |||
| AF069501.1 | AAC68867.1 | |||
| AF096276.1 | AAD19895.1 | |||
| AK077460.1 | ||||
| AK149445.1 | BAE28881.1 | |||
| AK165339.1 | ||||
| AK220310.1 | 1 | 505 | BAD90387.1 | |
| BC145696.1 |
More Information
Set Membership: |
|

Genomic Map