Assay Details
Target Gene Details
Entrez Gene ID: | 320910 |
Gene Name: | integrin beta 8 |
Gene Aliases: |
4832412O06Rik |
Location: |
Chr.12:119158186-119238276 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgb8 | NM_177290.3 | NP_796264.2 | ||
| AK029281.1 | ||||
| AK034644.1 | ||||
| BC120691.1 | ||||
| BC125343.1 |
More Information
Set Membership: |
|

Genomic Map