Assay Details
Target Gene Details
Entrez Gene ID: | 26423 |
Gene Name: | nuclear receptor subfamily 5, group A, member 1 |
Gene Aliases: |
Ad4BP, ELP, ELP-3, Ftz-F1, Ftzf1, SF-1, SF1 |
Location: |
Chr.2:38692656-38714560 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr5a1 | NM_139051.3 | NP_620639.1 | ||
| AB000490.1 | BAA19127.1 | |||
| AF511594.1 | AAM43952.1 | |||
| AK134378.1 | BAE22121.1 | |||
| AK144376.1 | BAE25855.1 | |||
| AK156911.1 | BAE33893.1 | |||
| BC110476.1 | AAI10477.1 | |||
| BC110477.1 | AAI10478.1 | |||
| D10584.1 | BAA01441.1 |
More Information
Set Membership: |
|

Genomic Map