Assay Details
Target Gene Details
Entrez Gene ID: | 73344 |
Gene Name: | RIKEN cDNA 1700034J05 gene |
Gene Aliases: |
- |
Location: |
Chr.6:146951301-146955058 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700034J05Rik | NM_001164236.1 | NP_001157708.1 | ||
| NM_028509.1 | NP_082785.1 | |||
| AK006598.1 | BAB24668.1 | |||
| AK076643.1 | BAC36432.1 | |||
| BC052362.1 | AAH52362.1 |
Target Gene Details
Entrez Gene ID: | 67533 |
Gene Name: | PTPRF interacting protein, binding protein 1 (liprin beta 1) |
Gene Aliases: |
4632409B19Rik, AW214454, AW261454 |
Location: |
Chr.6:146888080-147032025 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppfibp1 | NM_001170433.1 | NP_001163904.1 | ||
| NM_026221.2 | NP_080497.1 | |||
| AK146575.1 | BAE27271.1 | |||
| AK154482.1 | BAE32618.1 | |||
| AK173133.1 | BAD32411.1 | |||
| BC072603.1 | AAH72603.1 |
More Information
Set Membership: |
|

Genomic Map