Assay Details
Target Gene Details
Entrez Gene ID: | 19265 |
Gene Name: | protein tyrosine phosphatase, receptor type, C polypeptide-associated protein |
Gene Aliases: |
AI605264, CD-45-AP, LPAP, LSM-1 |
Location: |
Chr.19:4154646-4156710 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprcap | NM_016933.3 | 2 | 721 | NP_058629.2 |
| AI323196.1 | 2 | 693 | ||
| AI627045.1 | 1 | 356 | ||
| AK088334.1 | 2 | 719 | BAC40289.1 | |
| AK165203.1 | 2 | 719 | BAE38076.1 | |
| AW049152.1 | 1 | 142 | ||
| BC013273.1 | 2 | 720 | AAH13273.1 | |
| BC100560.1 | 2 | 702 | AAI00561.1 | |
| D10105.1 | 2 | 697 | BAA00986.1 | |
| U03856.1 | 1 | 714 | AAA67166.1 |

Genomic Map