Assay Details
Target Gene Details
Entrez Gene ID: | 22348 |
Gene Name: | solute carrier family 32 (GABA vesicular transporter), member 1 |
Gene Aliases: |
R75019, VGAT, Viaat |
Location: |
Chr.2:158610758-158615747 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc32a1 | NM_009508.2 | NP_033534.2 | ||
| AJ001598.1 | CAA04864.1 | |||
| AK140468.1 | BAE24402.1 | |||
| AK163661.1 | BAE37446.1 | |||
| AK163754.1 | BAE37482.1 | |||
| AY578289.1 | ||||
| BC052020.1 | AAH52020.1 |
More Information
Set Membership: |
|

Genomic Map