Assay Details
Target Gene Details
Entrez Gene ID: | 11770 |
Gene Name: | fatty acid binding protein 4, adipocyte |
Gene Aliases: |
422/aP2, ALBP/Ap2, Ap2, Lbpl |
Location: |
Chr.3:10204342-10208597 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fabp4 | NM_024406.2 | NP_077717.1 | ||
| AK003143.1 | BAB22601.1 | |||
| AK088535.1 | BAC40410.1 | |||
| BC054426.1 | AAH54426.1 | |||
| BY081693.1 | ||||
| CT010390.1 | CAJ18597.1 | |||
| K02109.1 | AAA39416.1 | |||
| M28726.1 | AAA37112.1 |
More Information
Set Membership: |
|

Genomic Map