Assay Details
Target Gene Details
Entrez Gene ID: | 68055 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F0 complex, subunit S |
Gene Aliases: |
1110015E18Rik, ATPW |
Location: |
Chr.12:69724930-69744665 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp5s | NM_026536.1 | NP_080812.1 | ||
| AK003725.1 | BAB22960.1 | |||
| AK009141.1 | ||||
| BC027442.1 | AAH27442.1 |
More Information
Set Membership: |
|

Genomic Map