Assay Details
Target Gene Details
Entrez Gene ID: | 19265 |
Gene Name: | protein tyrosine phosphatase, receptor type, C polypeptide-associated protein |
Gene Aliases: |
AI605264, CD-45-AP, LPAP, LSM-1 |
Location: |
Chr.19:4154646-4156710 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprcap | NM_016933.3 | NP_058629.2 | ||
| AA265746.1 | ||||
| AI323196.1 | ||||
| AK088334.1 | BAC40289.1 | |||
| AK165203.1 | BAE38076.1 | |||
| BB840651.1 | ||||
| BC013273.1 | AAH13273.1 | |||
| BC100560.1 | AAI00561.1 | |||
| D10105.1 | BAA00986.1 |

Genomic Map