Assay Details
Target Gene Details
Entrez Gene ID: | 71685 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14 |
Gene Aliases: |
0610033M06Rik |
Location: |
Chr.17:73493207-73710461 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt14 | NM_027864.2 | NP_082140.2 | ||
| AB175681.1 | BAD52069.1 | |||
| AK002747.1 | BAB22325.1 | |||
| AK049598.1 | ||||
| AK078292.1 | BAC37208.1 | |||
| AK090146.1 | ||||
| BC117800.1 | ||||
| BY733448.1 |
More Information
Set Membership: |
|

Genomic Map