Assay Details
Target Gene Details
Entrez Gene ID: | 56787 |
Gene Name: | achaete-scute complex homolog 3 (Drosophila) |
Gene Aliases: |
Sgn1, bHLHa42 |
Location: |
Chr.7:109727465-109731732 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ascl3 | NM_020051.1 | NP_064435.1 | ||
| AB046448.1 | BAB83911.1 | |||
| AJ277605.1 | CAC37689.1 | |||
| BC104346.1 | AAI04347.1 | |||
| BC104347.1 | AAI04348.1 |
More Information
Set Membership: |
|

Genomic Map