Assay Details
Target Gene Details
Entrez Gene ID: | 228839 |
Gene Name: | TGFB-induced factor homeobox 2 |
Gene Aliases: |
4921501K24, 5730599O09Rik, C80753, C81206 |
Location: |
Chr.2:156839817-156855575 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgif2 | NM_001291124.1 | NP_001278053.1 | ||
| NM_173396.3 | NP_775572.1 | |||
| AK017884.1 | ||||
| AK029490.1 | BAC26473.1 | |||
| AK157623.1 | BAE34136.1 | |||
| BC138705.1 | ||||
| BC138706.1 |
More Information
Set Membership: |
|

Genomic Map