Assay Details
Target Gene Details
Entrez Gene ID: | 15482 |
Gene Name: | heat shock protein 1-like |
Gene Aliases: |
Hsc70t, Msh5 |
Location: |
Chr.17:34972703-34979228 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspa1l | NM_013558.2 | NP_038586.2 | ||
| BC129894.1 | ||||
| BC129895.1 | ||||
| BY099756.1 | ||||
| D85732.1 | BAA32522.1 |
More Information
Set Membership: |
|

Genomic Map