Assay Details
Target Gene Details
Entrez Gene ID: | 67130 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6 (B14) |
Gene Aliases: |
14kDa, 2700038D15Rik, B230217P19Rik |
Location: |
Chr.15:82350139-82354291 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufa6 | NM_025987.3 | NP_080263.1 | ||
| AK002749.1 | BAB22326.1 | |||
| AK010988.1 | BAB27308.1 | |||
| AK012328.1 | BAB28165.1 | |||
| BC048451.1 | AAH48451.1 |
More Information
Set Membership: |
|

Genomic Map