Assay Details
Target Gene Details
Entrez Gene ID: | 50913 |
Gene Name: | oligodendrocyte transcription factor 2 |
Gene Aliases: |
AI604895, Bhlhb1, Olg-2, Oligo2, RK17, bHLHe19 |
Location: |
Chr.16:91225550-91228677 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olig2 | NM_016967.2 | NP_058663.2 | ||
| AB038697.1 | BAB18907.1 | |||
| AK080615.1 | BAC37958.1 | |||
| BC051967.1 | AAH51967.1 |
More Information
Set Membership: |
|

Genomic Map