Assay Details
Target Gene Details
Gene Symbol: | AmpR |
Gene Name: | Staphylococcus aureus beta-lactamase |
Gene Aliases: |
ApR, bla, blaZ, Amp |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gene Symbol: | AmpR |
Gene Name: | Staphylococcus aureus beta-lactamase |
Gene Aliases: |
ApR, bla, blaZ, Amp |