Assay Details
Target Gene Details
Gene Symbol: | HygR |
Gene Name: | Escherichia coli plasmid hygromycin B phosphotransferase |
Gene Aliases: |
HmB, HygroR, hgh, hph, hpt, Hyg |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gene Symbol: | HygR |
Gene Name: | Escherichia coli plasmid hygromycin B phosphotransferase |
Gene Aliases: |
HmB, HygroR, hgh, hph, hpt, Hyg |