Assay Details
Target Gene Details
Gene Symbol: | BsdR |
Gene Name: | Aspergillus terreus Blasticidin deaminase |
Gene Aliases: |
BSD, Bsd, G418-resistant |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gene Symbol: | BsdR |
Gene Name: | Aspergillus terreus Blasticidin deaminase |
Gene Aliases: |
BSD, Bsd, G418-resistant |