Assay Details
Target Gene Details
Entrez Gene ID: | 3075 |
Gene Name: | complement factor H |
Gene Aliases: |
AHUS1, AMBP1, ARMD4, ARMS1, CFHL3, FH, FHL1, HF, HF1, HF2, HUS |
Location: |
Chr.1:196651878-196747504 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFH | NM_000186.3 | NP_000177.2 | ||
| NM_001014975.2 | NP_001014975.1 | |||
| XM_017001108.1 | XP_016856597.1 | |||
| AK225649.1 | ||||
| AK226113.1 | ||||
| AK291395.1 | ||||
| BC037285.2 | AAH37285.1 | |||
| BC110643.1 | AAI10644.1 | |||
| BC142699.1 | ||||
| M12383.1 | AAA52013.1 | |||
| X04697.1 | CAB41739.1 | |||
| X07523.1 | CAA30403.1 | |||
| Y00716.1 | CAA68704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758990 | Chr.1:196621707 - 197200642 on Build GRCh38 | Gain+Loss |
|
| nsv548743 | Chr.1:196679339 - 196682935 on Build GRCh38 | Loss |
|
| nsv528460 | Chr.1:196573437 - 197477572 on Build GRCh38 | Gain |
|
| nsv818733 | Chr.1:196673103 - 196691131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map