Assay Details
Target Gene Details
Entrez Gene ID: | 9406 |
Gene Name: | zinc finger RANBP2-type containing 2 |
Gene Aliases: |
ZIS, ZIS1, ZIS2, ZNF265 |
Location: |
Chr.1:71063291-71081289 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZRANB2 | NM_005455.4 | 11 | 1899 | NP_005446.2 |
| NM_203350.2 | 10 | 1824 | NP_976225.1 | |
| AB209568.1 | 10 | 2635 | BAD92805.1 | |
| AF065391.1 | 10 | 1563 | AAD09746.1 | |
| AK023252.1 | 8 | 1378 | ||
| AK055983.1 | 2 | 1194 | ||
| AK123114.1 | 1 | 1151 | ||
| AK222858.1 | 10 | 1549 | BAD96578.1 | |
| AL136945.1 | 11 | 1621 | CAB66879.1 | |
| BC039814.1 | 11 | 1645 | AAH39814.1 | |
| BX538141.1 | 2 | 2437 | ||
| CR749325.1 | 1 | 4216 | CAH18180.1 | |
| CR749665.1 | 2 | 2437 | CAH18456.1 |
Target Gene Details
Entrez Gene ID: | 100132618 |
Gene Name: | ZRANB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:71046506-71067184 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZRANB2-AS1 | NR_038420.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546472 | Chr.1:70967301 - 71075320 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map