Assay Details
Target Gene Details
Entrez Gene ID: | 92745 |
Gene Name: | solute carrier family 38 member 5 |
Gene Aliases: |
JM24, SN2, SNAT5, pp7194 |
Location: |
Chr.X:48458537-48470256 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC38A5 | NM_033518.3 | NP_277053.2 | ||
| XM_005272694.3 | XP_005272751.2 | |||
| XM_005272695.4 | XP_005272752.3 | |||
| XM_005272697.2 | XP_005272754.2 | |||
| XM_005272698.4 | XP_005272755.2 | |||
| XM_006724569.3 | XP_006724632.1 | |||
| XM_017029960.1 | XP_016885449.1 | |||
| XM_017029961.1 | XP_016885450.1 | |||
| AF276889.1 | AAK61856.1 | |||
| AF318358.1 | AAL55865.1 | |||
| AK094788.1 | ||||
| BC019246.2 | AAH19246.1 | |||
| BC027721.2 | AAH27721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map