Assay Details
Target Gene Details
Entrez Gene ID: | 6419 |
Gene Name: | SET domain and mariner transposase fusion gene |
Gene Aliases: |
METNASE, Mar1 |
Location: |
Chr.3:4303304-4320649 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETMAR | NM_001243723.1 | 1 | 90 | NP_001230652.1 |
| NM_001276325.1 | 1 | 90 | NP_001263254.1 | |
| NM_001320676.1 | 1 | 90 | NP_001307605.1 | |
| NM_001320677.1 | 1 | 90 | NP_001307606.1 | |
| NM_001320678.1 | 1 | 90 | NP_001307607.1 | |
| NM_006515.3 | 1 | 90 | NP_006506.3 | |
| AF054989.1 | AAC09350.1 | |||
| AK222734.1 | BAD96454.1 | |||
| AK297865.1 | ||||
| AK302296.1 | ||||
| AK307537.1 | 1 | 90 | ||
| AY952295.1 | AAY29570.1 | |||
| BC008931.2 | AAH08931.2 | |||
| BC011635.2 | AAH11635.1 | |||
| DC357038.1 | 1 | 90 | ||
| U80776.1 | AAC52012.1 |
Target Gene Details
Entrez Gene ID: | 285362 |
Gene Name: | sulfatase modifying factor 1 |
Gene Aliases: |
AAPA3037, FGE, UNQ3037 |
Location: |
Chr.3:4034714-4467282 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUMF1 | XM_011533624.2 | XP_011531926.1 | ||
| XM_017006252.1 | XP_016861741.1 | |||
| XM_017006253.1 | XP_016861742.1 | |||
| XM_017006254.1 | XP_016861743.1 | |||
| AY358092.1 | AAQ88459.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575535 | Chr.3:4292219 - 4362841 on Build GRCh38 | Gain |
|
| dgv272n21 | Chr.3:4080585 - 4319799 on Build GRCh38 | Loss |
|
| esv3893640 | Chr.3:4196512 - 4421324 on Build GRCh38 | Loss |
|
| nsv1006124 | Chr.3:4235682 - 4310067 on Build GRCh38 | Loss |
|
| esv2759126 | Chr.3:4096685 - 4378673 on Build GRCh38 | Loss |
|
| nsv469899 | Chr.3:4281224 - 4315301 on Build GRCh38 | Loss |
|
| nsv589451 | Chr.3:4231805 - 4473507 on Build GRCh38 | Loss |
|
| dgv8185n54 | Chr.3:4183047 - 4304082 on Build GRCh38 | Loss |
|
| nsv997886 | Chr.3:4294748 - 4373592 on Build GRCh38 | Gain |
|
| nsv1003234 | Chr.3:4298679 - 4344910 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map