Assay Details
Target Gene Details
Entrez Gene ID: | 90799 |
Gene Name: | centrosomal protein 95 |
Gene Aliases: |
CCDC45 |
Location: |
Chr.17:64506736-64537951 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP95 | NM_001316990.1 | NP_001303919.1 | ||
| NM_138363.2 | NP_612372.1 | |||
| NR_133644.1 | ||||
| XM_005257779.3 | XP_005257836.1 | |||
| XM_005257780.2 | XP_005257837.1 | |||
| XM_006722166.3 | XP_006722229.1 | |||
| XM_011525458.2 | XP_011523760.1 | |||
| XM_017025287.1 | XP_016880776.1 | |||
| XM_017025288.1 | XP_016880777.1 | |||
| AK057326.1 | BAB71428.1 | |||
| AK290587.1 | ||||
| AK297409.1 | ||||
| AK316257.1 | ||||
| AL832822.1 | CAI46165.1 | |||
| BC009518.1 | AAH09518.1 | |||
| BP357015.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819879 | Chr.17:64508426 - 64508655 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map