Assay Details
Target Gene Details
Entrez Gene ID: | 9196 |
Gene Name: | potassium voltage-gated channel subfamily A regulatory beta subunit 3 |
Gene Aliases: |
AKR6A9, KCNA3.1B, KCNA3B, KV-BETA-3 |
Location: |
Chr.17:7921859-7929435 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNAB3 | NM_004732.3 | NP_004723.2 | ||
| XM_011524068.1 | 1 | 84 | XP_011522370.1 | |
| XM_017025304.1 | 1 | 84 | XP_016880793.1 | |
| XM_017025305.1 | 1 | 84 | XP_016880794.1 | |
| XM_017025306.1 | 1 | 84 | XP_016880795.1 | |
| AF016411.1 | AAB92499.1 | |||
| AK309378.1 | 1 | 611 | ||
| AK309807.1 | 1 | 603 | ||
| BC096232.1 | AAH96232.1 | |||
| BC096234.3 | AAH96234.1 | |||
| BC099634.1 | AAH99634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2670156 | Chr.17:7928627 - 7932174 on Build GRCh38 | Deletion |
|
| nsv574333 | Chr.17:7880977 - 7952801 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map